9-(4-Chloro-6-phenyl-1,3,5-triazin-2-yl)-9H-carbazole 97%

Supplier: Ambeed
Warning

A1144182-1G A1144182-25G A1144182-5G
76673-138EA 13.8 USD
76673-138 76673-140 76673-142
9-(4-Chloro-6-phenyl-1,3,5-triazin-2-yl)-9H-carbazole 97%
9-(4-Chloro-6-phenyl-1,3,5-triazin-2-yl)-9H-carbazole

Precaución: for Research Use Only

Formula: C₂₁H₁₃ClN₄
MW: 356.8077 g/mol
MDL Number: MFCD32206908
CAS Number: 1268244-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR