2,5-Bis(2-ethylhexyl)-3,6-di(furan-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 97%

Supplier: Ambeed
Warning

A1144088-1G A1144088-100MG A1144088-250MG
76673-120EA 108 USD
76673-120 77865-851 77865-852
2,5-Bis(2-ethylhexyl)-3,6-di(furan-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 97%
2,5-Bis(2-ethylhexyl)-3,6-di(furan-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione

Precaución: for Research Use Only

Formula: C₃₀H₄₀N₂O₄
MDL Number: MFCD31631763
CAS Number: 1286755-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR