2-((Difluoromethyl)sulfonyl)benzo[d]thiazole 97%

Supplier: Ambeed
Warning

A1144000-1G A1144000-250MG A1144000-5G A1144000-100MG
76673-106EA 59.42 USD
76673-106 76673-108 77570-804 77619-956
2-((Difluoromethyl)sulfonyl)benzo[d]thiazole 97%
2-((Difluoromethyl)sulfonyl)benzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₅F₂NO₂S₂
MW: 249.2576 g/mol
MDL Number: MFCD01831028
CAS Number: 186204-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR