4,5-Difluoro-2-(trimethylsilyl)phenyl trifluoromethanesulfonate 97%

Supplier: Ambeed
Danger

A1143810-100MG A1143810-1G A1143810-250MG
76673-098EA 46 USD
76673-098 76673-100 76673-102
4,5-Difluoro-2-(trimethylsilyl)phenyl trifluoromethanesulfonate 97%
4,5-Difluoro-2-(trimethylsilyl)phenyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₅O₃SSi
MW: 334.3400 g/mol
MDL Number: MFCD28969390
CAS Number: 217813-00-8
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR