(S)-1-(5-Fluoro-2-iodophenyl)ethanol 97%

Supplier: Ambeed
Warning

A1143703-10G A1143703-1G A1143703-5G A1143703-25G
76673-092EA 43 USD
76673-092 76673-094 76673-096 77865-846
(S)-1-(5-Fluoro-2-iodophenyl)ethanol 97%
(S)-1-(5-Fluoro-2-iodophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₈FIO
MDL Number: MFCD30803240
CAS Number: 1454847-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR