Potassium 2-(1-hydroxypentyl)benzoate 97%
Supplier: Ambeed
|
Warning
|
A1143690-250MG
A1143690-5G
76673-090EA
19.1
USD
76673-090
77619-950
Potassium 2-(1-hydroxypentyl)benzoate 97%
Potassium 2-(1-hydroxypentyl)benzoate
Precaución: for Research Use Only
Formula:
C₁₂H₁₅KO₃ MW: 246.3440 g/mol |
CAS Number:
685886-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...