Bis(4-fluorobenzyl)amine hydrochloride 97%

Supplier: Ambeed
Warning

A1143544-1G A1143544-5G
76673-078EA 68.96 USD
76673-078 76673-080
Bis(4-fluorobenzyl)amine hydrochloride 97%
Bis(4-fluorobenzyl)amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₄ClF₂N
MW: 269.7175 g/mol
MDL Number: MFCD32856098
CAS Number: 646051-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR