2,4-Bis(phenylsulfonyl)phenol 98%

Supplier: Ambeed
Warning

A1143437-100G A1143437-25G A1143437-500G
76673-060EA 19 USD
76673-060 76673-062 76673-064
2,4-Bis(phenylsulfonyl)phenol 98%
2,4-Bis(phenylsulfonyl)phenol

Precaución: for Research Use Only

Formula: C₁₈H₁₄O₅S₂
MDL Number: MFCD29072767
CAS Number: 177325-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR