Methyl 4-methyl-1H-indole-3-carboxylate 98%

Supplier: Ambeed
Warning

A1143375-100MG A1143375-1G A1143375-250MG A1143375-5G
76673-052EA 20.16 USD
76673-052 76673-054 76673-056 76673-058
Methyl 4-methyl-1H-indole-3-carboxylate 98%
Methyl 4-methyl-1H-indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.2100 g/mol
MDL Number: MFCD09841614
CAS Number: 858515-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR