3-Bromo-1,1':3',1''-terphenyl 97%

Supplier: Ambeed
Warning

A1143327-1G A1143327-5G
76673-050EA 27.59 USD
76673-050 77595-782
3-Bromo-1,1':3',1''-terphenyl 97%
3-Bromo-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₃Br
MW: 309.1998 g/mol
MDL Number: MFCD20486760
CAS Number: 98905-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR