N-(1-((5-Cyanopyridin-2-yl)methyl)-1H-pyrazol-3-yl)-2-(4-(1-(trifluoromethyl)cyclopropyl)phenyl)acetamide 98%

Supplier: Ambeed
Warning

A1143293-10MG A1143293-1MG A1143293-25MG A1143293-5MG
76673-042EA 202 USD
76673-042 76673-044 76673-046 76673-048
N-(1-((5-Cyanopyridin-2-yl)methyl)-1H-pyrazol-3-yl)-2-(4-(1-(trifluoromethyl)cyclopropyl)phenyl)acetamide 98%
N-(1-((5-Cyanopyridin-2-yl)methyl)-1H-pyrazol-3-yl)-2-(4-(1-(trifluoromethyl)cyclopropyl)phenyl)acetamide

Precaución: for Research Use Only

Formula: C₂₂H₁₈F₃N₅O
MDL Number: MFCD31813695
CAS Number: 1838651-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR