Cesium Pivalate 98%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dimethylpropionic acid Cesium salt, Pivalic acid Cesium salt, Trimethylacetic acid Cesium salt, Cesium 2,2-Dimethylpropionate, Cesium trimethylacetate

A1143284-100G A1143284-10G A1143284-1G A1143284-25G A1143284-5G
76673-032EA 743 USD
76673-032 76673-034 76673-036 76673-038 76673-040
Cesium Pivalate 98%
Cesium Pivalate

Precaución: for Research Use Only

Formula: C₅H₉CsO₂
MDL Number: MFCD10567040
CAS Number: 20442-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR