5-(2-Amino-4-chloro-5-sulfamoylphenyl)-1H-tetrazole 98%

Supplier: Ambeed
Danger

Synonyms: 4-Amino-2-chloro-5-(1H-tetrazol-5-yl)benzenesulphonamide

A1143282-25G A1143282-5G
76673-028EA 123.07 USD
76673-028 76673-030
5-(2-Amino-4-chloro-5-sulfamoylphenyl)-1H-tetrazole 98%
5-(2-Amino-4-chloro-5-sulfamoylphenyl)-1H-tetrazole

Precaución: for Research Use Only

Formula: C₇H₇ClN₆O₂S
MW: 274.68 g/mol
MDL Number: MFCD11870108
CAS Number: 82212-14-4
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR