Sodium dehydracetate monohydrate 99%

Supplier: Ambeed
Warning

Synonyms: Sodium 3-Acetyl-4-hydroxy-6-methyl-2-oxo-4(2H)-pyranolate, Sodium Dehydroacetate monohydrate, Sodium dehydroacetate monohydrate, Dehydroacetic acid sodium salt monohydrate, Sodium 1-(3,4-dihydro-6-methyl-2,4-dioxo-2H-pyran-3-ylidene)ethonolate monohydrate, 3-Acetyl-4-hydroxy-6-methyl-2-pyrone sodium salt, Sodium Dehydroacetate Monohydrate, Dehydracetic acid sodium salt monohydrate

A1143268-100G A1143268-500G
76673-024EA 21 USD
76673-024 76673-026
Sodium dehydracetate monohydrate 99%
Sodium dehydracetate monohydrate

Precaución: for Research Use Only

Formula: C₈H₇NaO₄·1H₂O
MDL Number: MFCD00040583
CAS Number: 64039-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR