Guanosine 5'-phosphate disodium salt hydrate 97%

Supplier: Ambeed
Warning

A1143216-100G A1143216-25G A1143216-500G
76673-012EA 36 USD
76673-012 76673-014 76673-016
Guanosine 5'-phosphate disodium salt hydrate 97%
Guanosine 5'-phosphate disodium salt hydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₅Na₂O₉P
MW: 425.1996 g/mol
MDL Number: MFCD00151238
CAS Number: 146894-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR