N4-(2,6-Dimethoxy-4-methyl-5-(3-(trifluoromethyl)phenoxy)quinolin-8-yl)pentane-1,4-diamine succinate(1:1) 98%

Supplier: Ambeed
Warning

Synonyms: Tafenoquine Succinate

A1143148-10MG A1143148-1MG A1143148-5MG
76673-006EA 218 USD
76673-006 76673-008 76673-010
N4-(2,6-Dimethoxy-4-methyl-5-(3-(trifluoromethyl)phenoxy)quinolin-8-yl)pentane-1,4-diamine succinate(1:1) 98%
N4-(2,6-Dimethoxy-4-methyl-5-(3-(trifluoromethyl)phenoxy)quinolin-8-yl)pentane-1,4-diamine succinate(1:1)

Precaución: for Research Use Only

Formula: C₂₈H₃₄F₃N₃O₇
MW: 581.5807 g/mol
MDL Number: MFCD00892249
CAS Number: 106635-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR