4,4',4''-Nitrilotribenzonitrile 97%

Supplier: Ambeed
Warning

A1143094-100MG A1143094-1G A1143094-250MG A1143094-5G
76672-998EA 19 USD
76672-998 76673-000 76673-002 76673-004
4,4',4''-Nitrilotribenzonitrile 97%
4,4',4''-Nitrilotribenzonitrile

Precaución: for Research Use Only

Formula: C₂₁H₁₂N₄
MDL Number: MFCD20265033
CAS Number: 51545-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR