O-Pivaloylhydroxylamine trifluoromethanesulfonate 97%

Supplier: Ambeed
Danger

A1143014-100G A1143014-1G A1143014-25G A1143014-5G
76672-982EA 1185 USD
76672-982 76672-984 76672-986 76672-988
O-Pivaloylhydroxylamine trifluoromethanesulfonate 97%
O-Pivaloylhydroxylamine trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₆H₁₂F₃NO₅S
MW: 267.2234 g/mol
MDL Number: MFCD32701977
CAS Number: 1293990-73-4
UN: 2925
ADR: 4.1,8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR