7-Bromo-3-oxoisoindoline-4-carbonitrile 98%

Supplier: Ambeed
Warning

A1143006-100MG A1143006-250MG
76672-978EA 36 USD
76672-978 76672-980
7-Bromo-3-oxoisoindoline-4-carbonitrile 98%
7-Bromo-3-oxoisoindoline-4-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅BrN₂O
MDL Number: MFCD27964212
CAS Number: 1370467-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR