6-Bromo-N-(3,4,5-trimethoxyphenyl)quinolin-4-amine 98%

Supplier: Ambeed
Warning

A1142890-10MG A1142890-1MG A1142890-5MG
76672-972EA 445.58 USD
76672-972 76672-974 76672-976
6-Bromo-N-(3,4,5-trimethoxyphenyl)quinolin-4-amine 98%
6-Bromo-N-(3,4,5-trimethoxyphenyl)quinolin-4-amine

Precaución: for Research Use Only

Formula: C₁₈H₁₇BrN₂O₃
MW: 389.24 g/mol
MDL Number: MFCD31689258
CAS Number: 2226517-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR