(2S,4R)-4-Hydroxy-1-((S)-3-methyl-2-(1-oxoisoindolin-2-yl)butanoyl)-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: VL285

A1142888-10MG A1142888-1MG A1142888-50MG A1142888-5MG A1142888-100MG A1142888-1G A1142888-250MG
76672-964EA 245.07 USD
76672-964 76672-966 76672-968 76672-970 77595-772 77595-774 77595-776
(2S,4R)-4-Hydroxy-1-((S)-3-methyl-2-(1-oxoisoindolin-2-yl)butanoyl)-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide 98%
(2S,4R)-4-Hydroxy-1-((S)-3-methyl-2-(1-oxoisoindolin-2-yl)butanoyl)-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₉H₃₂N₄O₄S
MW: 532.65 g/mol
MDL Number: MFCD32062670
CAS Number: 1448188-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR