RP-54745 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-5-(1-methyl-3,4-dihydroisoquinolin-2(1H)-yl)-3H-1,2-dithiol-3-one

A1142884-10MG A1142884-1MG A1142884-25MG A1142884-5MG
76672-956EA 46.68 USD
76672-956 76672-958 76672-960 76672-962
RP-54745 98%
RP-54745

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNOS₂
MW: 297.8235 g/mol
MDL Number: MFCD00879121
CAS Number: 135330-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR