2-(4-(4-Fluorobenzyl)piperidin-1-yl)-2-oxo-N-(2-oxo-2,3-dihydrobenzo[d]oxazol-6-yl)acetamide 98%

Supplier: Ambeed
Danger

Synonyms: Radiprodil

A1142878-10MG A1142878-1MG A1142878-25MG A1142878-50MG A1142878-5MG
76672-938EA 58.35 USD
76672-938 76672-940 76672-942 76672-944 76672-946
2-(4-(4-Fluorobenzyl)piperidin-1-yl)-2-oxo-N-(2-oxo-2,3-dihydrobenzo[d]oxazol-6-yl)acetamide 98%
2-(4-(4-Fluorobenzyl)piperidin-1-yl)-2-oxo-N-(2-oxo-2,3-dihydrobenzo[d]oxazol-6-yl)acetamide

Precaución: for Research Use Only

Formula: C₂₁H₂₀FN₃O₄
MW: 397.4 g/mol
MDL Number: MFCD11040157
CAS Number: 496054-87-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR