BAY-8002 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Chloro-5-(phenylsulphonyl)benzamido)benzoic acid

A1142862-10MG A1142862-1MG A1142862-25MG A1142862-50MG A1142862-5MG
76672-922EA 169.75 USD
76672-922 76672-924 76672-926 76672-928 76672-930
BAY-8002 98%
BAY-8002

Precaución: for Research Use Only

Formula: C₂₀H₁₄ClNO₅S
MW: 415.85 g/mol
MDL Number: MFCD05157149
CAS Number: 724440-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR