N-(3-((4-((3-(Trifluoromethyl)phenyl)amino)pyrimidin-2-yl)amino)phenyl)cyclopropanecarboxamide 98%
Supplier: Ambeed
|
Warning
|
A1142860-10MG
A1142860-1MG
A1142860-5MG
76672-916EA
50.93
USD
76672-916
76672-918
76672-920
N-(3-((4-((3-(Trifluoromethyl)phenyl)amino)pyrimidin-2-yl)amino)phenyl)cyclopropanecarboxamide 98%
N-(3-((4-((3-(Trifluoromethyl)phenyl)amino)pyrimidin-2-yl)amino)phenyl)cyclopropanecarboxamide
Precaución: for Research Use Only
Formula:
C₂₁H₁₈F₃N₅O MW: 413.4 g/mol |
MDL Number:
MFCD09265250 CAS Number: 879127-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...