2,3-Dimethoxynaphthalene-1,4-dicarbaldehyde 98%

Supplier: Ambeed
Warning

A1142544-100MG A1142544-1G A1142544-250MG A1142544-5G
76672-890EA 48 USD
76672-890 76672-892 76672-894 76672-896
2,3-Dimethoxynaphthalene-1,4-dicarbaldehyde 98%
2,3-Dimethoxynaphthalene-1,4-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₄
MW: 244.24 g/mol
MDL Number: MFCD32012304
CAS Number: 868623-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR