N-(4-Hydroxycyclohexyl)methanesulfonamide 95%

Supplier: Ambeed
Warning

A1141646-1G A1141646-250MG
76672-884EA 221.73 USD
76672-884 76672-886
N-(4-Hydroxycyclohexyl)methanesulfonamide 95%
N-(4-Hydroxycyclohexyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₇H₁₅NO₃S
MW: 193.26 g/mol
MDL Number: MFCD12179278
CAS Number: 1157109-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR