(R)-2-((tert-Butoxycarbonyl)amino)-5-methoxy-5-oxopentanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Glu(γ-OMe)-OH

A1141631-1G A1141631-25G A1141631-5G
76672-878EA 10.61 USD
76672-878 76672-880 76672-882
(R)-2-((tert-Butoxycarbonyl)amino)-5-methoxy-5-oxopentanoic acid 97%
(R)-2-((tert-Butoxycarbonyl)amino)-5-methoxy-5-oxopentanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₆
MW: 261.2717 g/mol
MDL Number: MFCD00136803
CAS Number: 76379-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR