3-Amino-4,5-dimethoxybenzoic acid 95%

Supplier: Ambeed
Warning

A1141037-100MG A1141037-1G A1141037-250MG A1141037-5G
76672-856EA 7 USD
76672-856 76672-858 76672-860 76672-862
3-Amino-4,5-dimethoxybenzoic acid 95%
3-Amino-4,5-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄
MDL Number: MFCD17276595
CAS Number: 861556-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR