4-(Benzyloxy)pyridine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A1140792-100MG A1140792-1G A1140792-250MG
76672-844EA 37.14 USD
76672-844 76672-846 76672-848
4-(Benzyloxy)pyridine-2-carboxylic acid 95%
4-(Benzyloxy)pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₃
MW: 229.2313 g/mol
MDL Number: MFCD11181375
CAS Number: 100727-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR