1,1,1-Trifluoro-2-methylbutan-2-amine hydrochloride 95%

Supplier: Ambeed
Warning

A1140268-100MG A1140268-250MG
76672-834EA 178.24 USD
76672-834 76672-836
1,1,1-Trifluoro-2-methylbutan-2-amine hydrochloride 95%
1,1,1-Trifluoro-2-methylbutan-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁ClF₃N
MW: 177.5958 g/mol
MDL Number: MFCD20502042
CAS Number: 1354952-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR