tert-Butyl N-(5-amino-2,4-difluorophenyl)carbamate 95%

Supplier: Ambeed
Warning

A1139586-100MG A1139586-1G A1139586-250MG
76672-828EA 97.61 USD
76672-828 76672-830 76672-832
tert-Butyl N-(5-amino-2,4-difluorophenyl)carbamate 95%
tert-Butyl N-(5-amino-2,4-difluorophenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₄F₂N₂O₂
MW: 244.2300 g/mol
MDL Number: MFCD12432529
CAS Number: 179741-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR