(R)-4-Hydroxythiochroman 1,1-dioxide 95%

Supplier: Ambeed
Warning

A1139083-100MG A1139083-1G A1139083-250MG
76672-822EA 14 USD
76672-822 76672-824 76672-826
(R)-4-Hydroxythiochroman 1,1-dioxide 95%
(R)-4-Hydroxythiochroman 1,1-dioxide

Precaución: for Research Use Only

Formula: C₉H₁₀O₃S
MDL Number: MFCD14705939
CAS Number: 1308650-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR