2-[4-(Aminomethyl)piperidin-1-yl]acetonitrile dihydrochloride 95%

Supplier: Ambeed
Warning

A1137377-100MG A1137377-50MG A1137377-250MG
76672-808EA 221 USD
76672-808 76672-810 77865-814
2-[4-(Aminomethyl)piperidin-1-yl]acetonitrile dihydrochloride 95%
2-[4-(Aminomethyl)piperidin-1-yl]acetonitrile dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₇Cl₂N₃
MDL Number: MFCD20441556
CAS Number: 1354953-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR