2-Chloro-4-isopropylphenol 95%

Supplier: Ambeed
Warning

A1136688-100MG A1136688-1G A1136688-250MG
76672-802EA 31 USD
76672-802 76672-804 76672-806
2-Chloro-4-isopropylphenol 95%
2-Chloro-4-isopropylphenol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO
MDL Number: MFCD16997172
CAS Number: 51202-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR