N-Methyl-1H-pyrazole-3-carboxamide 95%

Supplier: Ambeed
Warning

A1136353-1G A1136353-250MG A1136353-5G
76672-798EA 224.92 USD
76672-798 76672-800 77570-744
N-Methyl-1H-pyrazole-3-carboxamide 95%
N-Methyl-1H-pyrazole-3-carboxamide

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.1286 g/mol
MDL Number: MFCD19597056
CAS Number: 701214-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR