[4-(Cyclopentyloxy)phenyl]boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: B-[4-(Cyclopentyloxy)phenyl]boronic acid

A1135077-100MG A1135077-1G A1135077-250MG
76672-776EA 97 USD
76672-776 76672-778 76672-780
[4-(Cyclopentyloxy)phenyl]boronic acid 97%
[4-(Cyclopentyloxy)phenyl]boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₃
MDL Number: MFCD08461671
CAS Number: 871830-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR