4,7-Difluoro-1H-indole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A1134620-1G A1134620-250MG A1134620-5G
76672-774EA 92.3 USD
76672-774 77619-900 77619-902
4,7-Difluoro-1H-indole-2-carboxylic acid 95%
4,7-Difluoro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅F₂NO₂
MW: 197.1383 g/mol
MDL Number: MFCD01863159
CAS Number: 247564-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR