5-Chloro-6-fluoro-1-benzofuran-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A1132077-100MG A1132077-1G A1132077-250MG
76672-754EA 118 USD
76672-754 76672-756 76672-758
5-Chloro-6-fluoro-1-benzofuran-2-carboxylic acid 95%
5-Chloro-6-fluoro-1-benzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₄ClFO₃
MDL Number: MFCD28644688
CAS Number: 1781859-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR