3-Chloro-5-(chlorosulfonyl)-4-hydroxybenzoic acid 95%

Supplier: Ambeed
Danger

A1131216-100MG A1131216-1G A1131216-250MG
76672-742EA 42.44 USD
76672-742 76672-744 76672-746
3-Chloro-5-(chlorosulfonyl)-4-hydroxybenzoic acid 95%
3-Chloro-5-(chlorosulfonyl)-4-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₅S
MW: 271.0747 g/mol
MDL Number: MFCD18839087
CAS Number: 1201663-81-1
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR