6-Nitro-1H-indole-3-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A1130126-250MG A1130126-1G
76672-734EA 24 USD
76672-734 77619-890
6-Nitro-1H-indole-3-sulfonyl chloride 95%
6-Nitro-1H-indole-3-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₄S
MDL Number: MFCD00451765
CAS Number: 132745-00-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR