7-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)heptanoic acid 95%

Supplier: Ambeed
Warning

A1129871-100MG A1129871-250MG
76672-726EA 46 USD
76672-726 76672-728
7-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)heptanoic acid 95%
7-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)heptanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₄
MDL Number: MFCD00941277
CAS Number: 90267-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR