2-(2,3-Dichlorophenyl)cyclopropan-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A1129536-100MG A1129536-1G A1129536-250MG
76672-716EA 83 USD
76672-716 76672-718 76672-720
2-(2,3-Dichlorophenyl)cyclopropan-1-amine hydrochloride 95%
2-(2,3-Dichlorophenyl)cyclopropan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₀Cl₃N
MDL Number: MFCD18785529
CAS Number: 1311314-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR