(7-Methoxynaphthalen-2-yl)methanol 95%

Supplier: Ambeed
Warning

A1129388-100MG A1129388-250MG A1129388-1G
76672-712EA 51.99 USD
76672-712 76672-714 77619-886
(7-Methoxynaphthalen-2-yl)methanol 95%
(7-Methoxynaphthalen-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₂
MW: 188.2225 g/mol
MDL Number: MFCD18412893
CAS Number: 5665-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR