6-(4-Bromophenyl)piperidin-2-one 95%

Supplier: Ambeed
Warning

A1129245-100MG A1129245-1G
76672-708EA 185.66 USD
76672-708 76672-710
6-(4-Bromophenyl)piperidin-2-one 95%
6-(4-Bromophenyl)piperidin-2-one

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrNO
MW: 254.1231 g/mol
MDL Number: MFCD31396866
CAS Number: 267880-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR