4-(3-Chlorophenyl)-1H-pyrrole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A1129023-100MG A1129023-1G A1129023-250MG
76672-696EA 199.45 USD
76672-696 76672-698 76672-700
4-(3-Chlorophenyl)-1H-pyrrole-2-carboxylic acid 95%
4-(3-Chlorophenyl)-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₈ClNO₂
MW: 221.6397 g/mol
MDL Number: MFCD26604420
CAS Number: 1511974-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR