3-(3-Hydroxyphenyl)-2-propynoic acid 95%

Supplier: Ambeed
Warning

A1128691-100MG A1128691-250MG
76672-692EA 53.05 USD
76672-692 76672-694
3-(3-Hydroxyphenyl)-2-propynoic acid 95%
3-(3-Hydroxyphenyl)-2-propynoic acid

Precaución: for Research Use Only

Formula: C₉H₆O₃
MW: 162.1400 g/mol
MDL Number: MFCD20639523
CAS Number: 10401-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR