2,6-Dichloroquinoline-4-carbonitrile 95%

Supplier: Ambeed
Warning

A1128023-100MG A1128023-250MG
76672-680EA 122.01 USD
76672-680 76672-682
2,6-Dichloroquinoline-4-carbonitrile 95%
2,6-Dichloroquinoline-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₄Cl₂N₂
MW: 223.0582 g/mol
MDL Number: MFCD18483205
CAS Number: 50504-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR