6-((Ethylamino)methyl)pyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A1127956-1G A1127956-250MG
76672-676EA 190 USD
76672-676 76672-678
6-((Ethylamino)methyl)pyridin-2(1H)-one 95%
6-((Ethylamino)methyl)pyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O
MDL Number: MFCD28153559
CAS Number: 1692781-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR