6-((tert-Butylamino)methyl)pyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A1127955-1G A1127955-250MG
76672-672EA 335.25 USD
76672-672 76672-674
6-((tert-Butylamino)methyl)pyridin-2(1H)-one 95%
6-((tert-Butylamino)methyl)pyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O
MW: 180.2468 g/mol
CAS Number: 1697774-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR